Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human APRIL Recombinant Protein, PeproTech®

Catalog No. 31010C2UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-10C-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 310-10C-2UG Supplier Gibco™ Supplier No. 31010C2UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

310-10C-01M will be provided as 20 x 50 μg (310-10C-50UG). 310-10C-500 will be provided as 10 x 50 μg (310-10C-50UG). 310-10C-250 will be provided as 5 x 50 μg (310-10C-50UG). 310-10C-100 will be provided as 2 x 50 μg (310-10C-50UG) Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues. *Human APRIL (Catalog Number 310-10C) has replaced Human APRIL (Catalog Number 310-10). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75888
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 8741
Molecular Weight (g/mol) 16.3 kDa
Name Human APRIL
Purification Method Purified
Quantity 2 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 2310026N09Rik; A proliferation-inducing ligand; APRIL; CD256; TALL2; TALL-2; TNF- and APOL-related leukocyte expressed ligand 2; TNF-related death ligand 1; Tnfsf13; TNLG7B; Trdl1; TRDL-1; tumor necrosis factor (ligand) superfamily, member 13; tumor necrosis factor ligand 7B; tumor necrosis factor ligand superfamily member 13; tumor necrosis factor superfamily member 13; tumor necrosis factor-like protein ZTNF2; tumor necrosis factor-related death ligand-1; UNQ383/PRO715; ZTNF2
Common Name APRIL
Gene Symbol TNFSF13
Biological Activity Determined by its ability to induce NF-kappaB activation in HEK293-NF-kappaB reporter cells transfected with BCMA receptor.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVLTQKQKKQ HSVLHLVPIN ATSKDDSDVT EVMWQPALRR GRGLQAQGYG VRIQDAGVYL LYSQVLFQDV TFTMGQVVSR EGQGRQETLF RCIRSMPSHP DRAYNSCYSA GVFHLHQGDI LSVIIPRARA KLNLSPHGTF LGFVKL
Content And Storage -20°C
Activity Determined by its ability to induce NF-kappaB activation in HEK293-NF-kappaB reporter cells transfected with BCMA receptor.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.