Learn More
Sino Biological Anti-AQP3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 205252T46100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human AQP3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1516