Learn More
Biomatik Corporation Human Aquaporin 5 (AQP5) ELISA Kit
Supplier:  Biomatik Corporation EKU10661
Encompass_Preferred
Human Aquaporin 5 (AQP5) ELISA Kit for detecting AQP5 in serum, plasma, saliva, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 246.9-20,000pg/ml. Sensitivity: 91.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P55064. Gene ID: 362. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5798