Learn More
American Research Products Inc Human Arachidonic Acid AA ELI
Catalog No. 5000326727
Encompass
Human Arachidonic Acid AA ELISA Kit from CUSABIO Detection Range 78 125 ng/mL-5000 ng/mL Sensitivity 69 748 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26727 Supplier American Research Products Inc Supplier No. CSBE09040H
Human Arachidonic Acid AA ELISA Kit from CUSABIO Detection Range 78 125 ng/mL-5000 ng/mL Sensitivity 69 748 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive