Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ARAP2 (aa 1614-1702) Control Fragment Recombinant Protein

Catalog No. RP96597
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96597 Supplier Invitrogen™ Supplier No. RP96597
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57465. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ARAP2 also known as Centaurin-delta-1 is a protein with ARF-GAP, RHO-GAP, ankyrin repeat, RAS-associating, and pleckstrin homology domains. A phosphatidylinositol-3, 4, 5-trisphosphate-dependent GTPase-activating protein, ARAP2, binds to RhoA-GTP, but lacks the predicted catalytic arginine in the RHO-GAP domain and does not have RHO-GAP activity. It modulates actin cytoskeleton remodeling by regulating ARF and RHO family members. Usually ARAP2 is activated by phosphatidylinositol-3, 4, 5-trisphosphate (PtdIns(3, 4, 5)P3) binding and can be activated by phosphatidylinositol-3, 4-bisphosphate (PtdIns(3, 4, 5)P2) binding, albeit with lower efficiency. ARAP2 associates with focal adhesions and functions downstream of RhoA to regulate focal adhesion dynamics. It is usually detected in brain, thymus, lymph node, thyroid, spinal cord, trachea, heart, skeletal muscle, spleen, kidney, liver, placenta, lung and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WZ64
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 116984
Name Human ARAP2 (aa 1614-1702) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Arap2; Arf and Rho GAP adapter protein 2; arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2; centaurin; centaurin, delta 1; centaurin-delta-1; CENTD1; Cnt-d1; Gm148; KIAA0580; mKIAA0580; PARX; PARX protein; Protein PARX
Common Name ARAP2
Gene Symbol ARAP2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HCLEHKDDKLRNRPRKHRSFNCLEDTEPEAPLGQPKGHKGLKTLRKTEDRNSKATLDSDHKLPSRVIEELNVVLQRSRTLPKELQDEQI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.