Learn More
Sino Biological Human ARFGAP1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 303ARAH0010250
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ARFGAP1, and purified by antigen affinity chromatography.
Catalog No. 50-237-4307