Learn More
Sino Biological Recombinant Human Arginase 1/ARG1 Protein (His Tag)
Supplier:  Sino Biological 11558H08H100
Encompass_Preferred
A DNA sequence encoding the human ARG1 isoform 1 (P05089-1) (Met 1-Lys 322) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6785