Learn More
Sino Biological Anti-Arginase 1/ARG1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11558RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ARG1 / Arginase 1 (rh ARG1 / Arginase 1; Catalog#11558-H08H; P05089-1; Met 1-Lys 322). ARG1 / Arginase 1 specific IgG was purified by Human ARG1 / Arginase 1 affinity chromatography.
Catalog No. 50-255-4689