Learn More
Abcam Human Arg2 ELISA Kit, 1 x 96 Tests
Catalog No. 502661856
Encompass_Preferred
Human Arg2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Arg2 with a sensitivity of 13.177 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-1856 Supplier Abcam Supplier No. AB2897021X96TESTS
Human Arg2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Arg2 with a sensitivity of 13.177 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement