Learn More
Biomatik Corporation Human Arginine Vasopressin Induced Protein 1 (AVPI1) ELISA Kit
Supplier:  Biomatik Corporation EKU12519
Encompass_Preferred
Human Arginine Vasopressin Induced Protein 1 (AVPI1) ELISA Kit for detecting AVPI1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5T686. Gene ID: 60370. Target Synonyms: VIP32; VIT32; AVP-induced protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5061