Learn More
Biomatik Corporation Human Argininosuccinate Synthetase 1 (ASS1) ELISA Kit
Supplier:  Biomatik Corporation EKU02569
Encompass_Preferred
Human Argininosuccinate Synthetase 1 (ASS1) ELISA Kit for detecting ASS1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00966. Gene ID: 445. Target Synonyms: CTLN1; Citrulline--aspartate ligase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9116