Learn More
Biomatik Corporation Human Arginyl tRNA Synthetase (RARS) ELISA Kit
Supplier:  Biomatik Corporation EKU02570
Encompass_Preferred
Human Arginyl tRNA Synthetase (RARS) ELISA Kit for detecting RARS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54136. Gene ID: 5917. Target Synonyms: ArgRS; DALRD1; Arginine tRNA Ligase 1,Cytoplasmic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6706