Learn More
Sino Biological Anti-ARHGAP11A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107604T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ARHGAP11A, and purified by antigen affinity chromatography.
Catalog No. 50-241-2267