Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Aromatase (aa 340-415) Control Fragment Recombinant Protein

Catalog No. RP101314
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101314 Supplier Invitrogen™ Supplier No. RP101314
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111329. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11511
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 1588
Name Human Aromatase (aa 340-415) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Ar; ArKO; ARO; ARO1; Arom; Aromatase; aromatase 1; aromatase cytochrome P450; aromatase cytochrome P450 19A1; CPV1; CYAR; Cyp19; Cyp19a; Cyp19a1; CYP19P1; CYPXIX; CYPXIXA1; Cytochrome P450 17-alpha; cytochrome P450 19 type I; Cytochrome P450 19A1; cytochrome P450 aromatase; cytochrome P-450 aromatase; cytochrome P-450 aromatase (alt.); cytochrome P450 family 19 subfamily a; cytochrome P450 family 19 subfamily A member 1; Cytochrome P450, 19, aromatase; cytochrome P450, family 17, subfamily A, polypeptide 1; cytochrome P450, family 19, subfamily A, polypeptide 1; cytochrome P450, family XIX (conversion of androgen to estrogen), aromatase; cytochrome P450, family19, subfamily A, polypeptide 1; cytochrome P450, subfamily 19; cytochrome P450, subfamily XIX (aromatization of androgens); cytochrome P-450AROM; estrogen synthase; estrogen synthetase; flavoprotein-linked monooxygenase; Int5; Int-5; MGC104309; microsomal monooxygenase; P450AROM; P-450AROM; type III cytochrome p450 aromatase
Common Name Aromatase
Gene Symbol Cyp19a1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IGERDIKIDDIQKLKVMENFIYESMRYQPVVDLVMRKALEDDVIDGYPVKKGTNIILNIGRMHRLEFFPKPNEFTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.