Learn More
Sino Biological Anti-ARSA Antibody (HRP), Rabbit Polyclonal
Supplier:  Sino Biological 10449RP02H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ARSA (rh ARSA; Catalog#10449-H08H; P15289-1; Met 1-Ala 507). ARSA specific IgG was purified by human ARSA affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2214