Learn More
Sino Biological Anti-ART3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13542T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ART3 (rh ART3; Catalog#13542-H08H; AAH08397.1; Met1-Ser362). ART3 specific IgG was purified by Human ART3 affinity chromatography.
Catalog No. 50-255-4221