Learn More
Sino Biological Human ART3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13542T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ART3 (rh ART3; Catalog#13542-H08H; AAH08397.1; Met1-Ser362). ART3 specific IgG was purified by Human ART3 affinity chromatography.
Catalog No. 50-237-3695