Learn More
Bioworld HUMAN ARTEMIN (RECOMBINANT)
Supplier:  Bioworld 220602491
Encompass_Preferred
Recombinant Human Artemin - ART Source: Baculovirus Description: Recombinant Human Artemin produced in E.Coli is a disulfide-linked homodimer, non-glycosylated, polypeptide chain containing 2 x 113 amino acids and having a total mole...
Catalog No. 50-253-347