Learn More
Biomatik Corporation Human Aryl Hydrocarbon Receptor (AhR) CLIA Kit
Supplier:  Biomatik Corporation EKU10026
Encompass_Preferred
Human Aryl Hydrocarbon Receptor (AhR) CLIA Kit for detecting AhR in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P35869. Gene ID: 196. Target Synonyms: BHLHE76; Class E basic helix-loop-helix protein 76. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7511