Learn More
Biomatik Corporation Human Arylalkylamine-N-Acetyltransferase (AANAT) ELISA Kit
Supplier:  Biomatik Corporation EKU09095
Encompass_Preferred
Human Arylalkylamine-N-Acetyltransferase (AANAT) ELISA Kit for detecting AANAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16613. Gene ID: 15. Target Synonyms: AA-NAT; SNAT; Aralkylamine N-Acetyltransferase; Serotonin Acetyltransferase; Serotonin Acetylase; Serotonin N-Acetyltransferase; Melatonin Rhythm Enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6464