Learn More
Biomatik Corporation Human Arylformamidase (AFMID) ELISA Kit
Supplier:  Biomatik Corporation EKU02582
Encompass_Preferred
Human Arylformamidase (AFMID) ELISA Kit for detecting AFMID in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63HM1. Gene ID: 125061. Target Synonyms: KF; KFA; FKF; Kynurenine formamidase; N-formylkynurenine formamidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5080