Learn More
Sino Biological Human ASF1B MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14382MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ASF1B (rh ASF1B; Catalog#14382-H08B; Q9NVP2; Met1-Ile202). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1981