Learn More
ACROBiosystems Biotinylated Human ASGR1 / ASGPR1 Trimer, His,Avitag (GS1-H82Q3) is expressed from human 293 cells (HEK293). It contains AA Gln 62 - Leu 291 (Accession # P07306-1).
Supplier:  ACROBiosystems GS1H82Q3200UG
Encompass_Preferred
ASGPR, a transmembrane C-type lectin, recognizes a wide variety of ligands that contain either terminal galactose (Gal) or N-acetylgalactosamine (GalNAc) residues and has been identified as a highly selective receptor on hepatocytes. The ASGR is composed of both a major (ASGR1) and minor subunit (ASGR2). ASGR1 has been shown to be efficiently targeted to the plasma membrane and to undergo constitutive endocytosis and recycling and found mainly expressed in the human liver.
Catalog No. 50-223-1089