Learn More
ACROBiosystems Human ASGR1/ASGPR1 Protein, Fc Tag
Supplier:  ACROBiosystems AS1H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human ASGR1, Fc Tag (AS1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 62 - Leu 291 (Accession # P07306-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 52.8 kDa.
Catalog No. 50-205-5012