Learn More
Sino Biological Recombinant Anti-ASGR1 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10773R024H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGPR1 (rh ASGPR1 ; Catalog#10773-H07H; P07306; Gln62-Leu291), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-7670