Learn More
Sino Biological Recombinant Anti-ASGR1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10773R023100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGPR1 (rh ASGPR1; Catalog#10773-H07H; P07306; Gln62-Leu291).
Catalog No. 50-240-7774