Learn More
Sino Biological Human ASGR1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10773R02120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGPR1 (rh ASGPR1 ; Catalog#10773-H07H; P07306; Gln62-Leu291).
Catalog No. 50-237-3256