Learn More
Sino Biological Anti-ASGR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10773T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ASGR1 (rh ASGR1; Catalog#10773-H07H; P07306; Gln62-Leu291). ASGR1 specific IgG was purified by Human ASGR1 affinity chromatography.
Catalog No. 50-255-5172