Learn More
Sino Biological Anti-ASGR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10773RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ASGPR1 (rh ASGR1; Catalog#10773-H07H; P07306; Gln62-Leu291). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8665