Learn More
ACROBiosystems HEK293/Human ASGR1&ASGR2 Stable Cell Line
Catalog No. 502719737
Encompass_Preferred
ASGPR, a transmembrane C-type lectin, recognizes a wide variety of ligands that contain either terminal galactose (Gal) or N-acetylgalactosamine (GalNAc) residues and has been identified as a highly selective receptor on hepatocytes. The ASGR is composed of both a major (ASGR1) and minor subunit (ASGR2).
Catalog No. 50-271-9737 Supplier ACROBiosystems Supplier No. CHEKATP1722VIALS
ASGPR, a transmembrane C-type lectin, recognizes a wide variety of ligands that contain either terminal galactose (Gal) or N-acetylgalactosamine (GalNAc) residues and has been identified as a highly selective receptor on hepatocytes. The ASGR is composed of both a major (ASGR1) and minor subunit (ASGR2).