Learn More
Sino Biological Recombinant Human ASGR2 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 13908H07H1
Encompass_Preferred
A DNA sequence encoding the human ASGR2 (P07307-1) (Gln80-Ala311) was expressed, with an N-terminal polyhistidine tag.
Catalog No. 50-224-7153