Learn More
BIOHIPPO Human ASGR2 Microsample
SDP
Catalog No. 5000315321
Encompass
Human ASGR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ASGR2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 27 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ASGR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ASGR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ASGR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15321 Supplier BIOHIPPO Supplier No. BHE15211803
Human ASGR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ASGR2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 27 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ASGR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ASGR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ASGR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color