Learn More
Sino Biological Anti-ASGR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13908T34100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ASGR2 (rh ASGR2; Catalog#13908-H07H; P07307-1; Gln80-Ala311). ASGR2 specific IgG was purified by Human ASGR2 affinity chromatography.
Catalog No. 50-255-4162