Learn More
Biomatik Corporation Human Asparagine Synthetase (ASNS) ELISA Kit
Supplier:  Biomatik Corporation EKU02589
Encompass_Preferred
Human Asparagine Synthetase (ASNS) ELISA Kit for detecting ASNS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08243. Gene ID: 440. Target Synonyms: Aspartate-Ammonia Ligase; Asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9114