Learn More
Abcam Human Aspartate Aminotransferase ELISA Kit, 96TEST
Supplier:  Abcam AB28529996TEST
Encompass_Preferred
Human Aspartate Aminotransferase ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Aspartate Aminotransferase in Human in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5478