Learn More
Biomatik Corporation Human Aspartate Beta Hydroxylase (ASPH) ELISA Kit
Supplier:  Biomatik Corporation EKU02596
Encompass_Preferred
Human Aspartate Beta Hydroxylase (ASPH) ELISA Kit for detecting ASPH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12797. Gene ID: 444. Target Synonyms: BAH; CASQ2BP1; HAAH; JCTN; Junctin; Humbug; Junctate; Aspartyl/Asparaginyl Beta-Hydroxylase; Peptide-aspartate beta-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7733