Learn More
Biomatik Corporation Human Asporin (ASPN) ELISA Kit
Supplier:  Biomatik Corporation EKU02597
Encompass_Preferred
Human Asporin (ASPN) ELISA Kit for detecting ASPN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BXN1. Gene ID: 54829. Target Synonyms: PLAP1; SLRR1C; Periodontal ligament-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7734