Learn More
Abcam Human AST ELISA Kit (Aspartate Aminotransferase), 1 x 96 Tests
Catalog No. 502656715
Encompass_Preferred
Human AST ELISA Kit (Aspartate Aminotransferase) is a single-wash 90-min Simplestep used to quantify Human AST (Aspartate Aminotransferase) with a sensitivity of 43 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6715 Supplier Abcam Supplier No. AB2638811X96TESTS
Human AST ELISA Kit (Aspartate Aminotransferase) is a single-wash 90-min Simplestep used to quantify Human AST (Aspartate Aminotransferase) with a sensitivity of 43 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement