Learn More
Sciencell Research Laboratories Human Astrocytes 1 x 10^6 cells in 1 ml volume.
Catalog No. NC9791361
Encompass
Isolated from human brain (cerebral cortex). HA are cryopreserved at passage one and delivered frozen. Each vial contains >1 x 10^6 cells in 1 ml volume. Storage: Directly and immediately transfer cells from dry ice to liquid nitrogen upon receiving and keep the cells in liquid nitrogen until cell culture is needed for experiments.
Catalog No. NC9791361 Supplier Sciencell Research Laboratories Supplier No. 1800
Isolated from human brain (cerebral cortex). HA are cryopreserved at passage one and delivered frozen. Each vial contains >1 x 10^6 cells in 1 ml volume. Storage: Directly and immediately transfer cells from dry ice to liquid nitrogen upon receiving and keep the cells in liquid nitrogen until cell culture is needed for experiments.