Learn More
ATCC Human astrovirus (HAstV) Type 2, ATCC VR-1943 - Frozen research strain
Supplier:  ATCC 502383896
Encompass
Human astrovirus (HAstV) type 2 is a frozen viral research strain used for infectious disease research, assay development, vaccine research, and enteric disease studies. Isolated from a human stool sample, this Mamastrovirus has a sequenced genome and is supplied frozen for ultralow-temperature storage.
  • Frozen viral culture format suitable for laboratory research.
  • Genome sequenced with supporting genomic data available.
  • Isolated from human stool (Oxford, United Kingdom).
  • Requires storage at -70°C or colder to maintain viability.
  • Common applications: infectious disease research, assay development, vaccine research.
  • For research use only; not for diagnostic or therapeutic use.
Catalog No. 50-238-3896