Learn More
Biomatik Corporation Human Ataxin 10 (ATXN10) ELISA Kit
Supplier:  Biomatik Corporation EKU08444
Encompass_Preferred
Human Ataxin 10 (ATXN10) ELISA Kit for detecting ATXN10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBB4. Gene ID: 25814. Target Synonyms: E46L; SCA10; Spinocerebellar Ataxia 10; Brain protein E46 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7739