Learn More
Sino Biological Anti-ATF2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11599T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ATF2 (rh ATF2; Catalog#11599-H20B; P15336-1; Met1-Ser505). ATF2 specific IgG was purified by Human ATF2 affinity chromatography.
Catalog No. 50-255-4684