Learn More
Raybiotech Inc C-Series Human Atherosclerosis Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHATH18
Encompass_Preferred
Detects 20 Human Atherosclerotic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1 (CCL11), GCSF, GDNF, GM-CSF, ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 R1, IL-1 R2, MCP-1 (CCL2), M-CSF, TGF beta 1, TGF beta 2, TGF beta 3, TNF alpha, TNF beta (TNFSF1B), MIP-3 alpha (CCL20), PDGF-BB, RANTES (CCL5)
Catalog No. 50-194-8488