Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter C11 (ABCC11) ELISA Kit
Supplier:  Biomatik Corporation EKU02610
Encompass_Preferred
Human ATP Binding Cassette Transporter C11 (ABCC11) ELISA Kit for detecting ABCC11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.103ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96J66. Gene ID: 85320. Target Synonyms: ABC-C11; MRP8; EWWD; Multidrug resistance-associated protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5283