Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter D1 (ABCD1) ELISA Kit
Supplier:  Biomatik Corporation EKU10669
Encompass_Preferred
Human ATP Binding Cassette Transporter D1 (ABCD1) ELISA Kit for detecting ABCD1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33897. Gene ID: 215. Target Synonyms: ABC-D1; ABC42; ALD; ALDP; AMN; Adrenoleukodystrophy protein; ATP-Binding Cassette,Subfamily D (ALD) Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5385