Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter C10 (ABCC10) ELISA Kit
Supplier:  Biomatik Corporation EKU02609
Encompass_Preferred
Human ATP Binding Cassette Transporter C10 (ABCC10) ELISA Kit for detecting ABCC10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5T3U5. Gene ID: 89845. Target Synonyms: MRP7; SIMRP7; Multidrug Resistance Associated Protein 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7124