Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter A4 (ABCA4) ELISA Kit
Supplier:  Biomatik Corporation EKU02608
Encompass_Preferred
Human ATP Binding Cassette Transporter A4 (ABCA4) ELISA Kit for detecting ABCA4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P78363. Gene ID: 24. Target Synonyms: ABC-A4; ABC10; ABCR; ARMD2; CORD3; FFM; RMP; RP19; STGD; STGD1; Stargardt disease protein; Retinal-specific ATP-binding cassette transporter; RIM ABC transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7455