Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter A7 (ABCA7) ELISA Kit
Supplier:  Biomatik Corporation EKU10668
Encompass_Preferred
Human ATP Binding Cassette Transporter A7 (ABCA7) ELISA Kit for detecting ABCA7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IZY2. Gene ID: 10347. Target Synonyms: ABC-A7; ABCA-SSN; ABCX; Autoantigen SS-N; Macrophage ABC transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5245