Learn More
Sino Biological Anti-ATP12A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103497T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP12A, and purified by antigen affinity chromatography.
Catalog No. 50-241-0766