Learn More
Sino Biological Anti-ATP1B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14255T40100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ATP1B1 (rh ATP1B1; Catalog#14255-H07H; P05026-1; Glu63-Ser303). ATP1B1 specific IgG was purified by Human ATP1B1 affinity chromatography.
Catalog No. 50-255-4141